Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Viral B8R Biotinylated Protein

Catalog No. BT8225025
Change view
Click to view available options
25μg; Biotin
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
BT8225025 25μg; Biotin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BT8225025 Supplier R&D Systems™ Supplier No. BT8225025
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Viral B8R Biotinylated Protein is derived from CHO. The Recombinant Viral B8R Biotinylated Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P24770
For Use With (Application) Bioactivity
Gene ID (Entrez) 3707661
Molecular Weight (g/mol) Theoretical M.W.: 30kDa (unlabeled)
Quantity 25μg
Source Chinese Hamster Ovary cell line,CHO-derived viral B8R protein Lys18-Ser272,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Common Name B8R
Gene Symbol B8R
Biological Activity Measured by its ability to inhibit rhIFN-gamma mediated protection of HeLa human cervical epithelial carcinoma cells to viral lysis. Meager, A. (1987) in Lymphokines and Interferons, a Practical Approach. Clemens, M.J. et al. (eds): IRL Press. 129. Th ED50 for this effect is typically 0.5-3ng/mL.
Conjugate Biotin
Species Virus
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Lys18-Ser272, with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.