Learn More
MSC PIP Recon Standard Safety Glasses, Polycarbonate, FogLess 3Sixty, Rimless, 1 ea
Catalog No. 5000688905
Encompass
These safety glasses feature a lightweight, rimless design with a 9 base curve polycarbonate lens, providing durable eye protection for construction and general purpose applications. Engineered with FogLess 3Sixty™ technology, the lenses offer enhanced hydrophobic action to resist fogging and ensure clear vision in extreme working conditions.
  • Anti-scratch and anti-fog lens coatings
  • Flexible bayonet-style temples with rubber pads for a secure, non-slip fit
  • Adjustable rubber nosepiece for enhanced comfort
  • Meets ANSI Z87.1+ and CSA Z94.3 standards
  • Light gray lens tint suitable for indoor and outdoor applications
Catalog No. 50-006-88905 Supplier MSC Supplier No. 19140474
These safety glasses feature a lightweight, rimless design with a 9 base curve polycarbonate lens, providing durable eye protection for construction and general purpose applications. Engineered with FogLess 3Sixty™ technology, the lenses offer enhanced hydrophobic action to resist fogging and ensure clear vision in extreme working conditions.
  • Anti-scratch and anti-fog lens coatings
  • Flexible bayonet-style temples with rubber pads for a secure, non-slip fit
  • Adjustable rubber nosepiece for enhanced comfort
  • Meets ANSI Z87.1+ and CSA Z94.3 standards
  • Light gray lens tint suitable for indoor and outdoor applications