Learn More
Med Vet International Reconstruction Plate, J0374P, 3.5mm, 7 Holes, 82mm, Each
Catalog No. 5000582277
Encompass_Preferred
JorVet is pleased to introduce a full range of cuttable plates from 1.5mm through to 3.5mm. The holes are round and spaced to offer the maximum number of screws per bone fragment. Plates are contoured longitudinally for extra strength. Simply cut off as many holes as you need. The 2.0mm plates are available as cuttable/malleable versions and function as easily contoured reconstruction plates. 3.5 mm Reconstruction PlatesHoles: 7Length in mm: 82
Catalog No. 50-005-82277 Supplier Med Vet International Supplier No. JOR374P
JorVet is pleased to introduce a full range of cuttable plates from 1.5mm through to 3.5mm. The holes are round and spaced to offer the maximum number of screws per bone fragment. Plates are contoured longitudinally for extra strength. Simply cut off as many holes as you need. The 2.0mm plates are available as cuttable/malleable versions and function as easily contoured reconstruction plates. 3.5 mm Reconstruction PlatesHoles: 7Length in mm: 82