Learn More
Med Vet International Reconstruction Plates - 2.7mm - 7 holes, J0374PS, Each
Catalog No. 5000582780
Encompass_Preferred
JorVet is pleased to introduce a full range of cuttable plates from 1.5mm through to 3.5mm. The holes are round and spaced to offer the maximum number of screws per bone fragment. Plates are contoured longitudinally for extra strength. Simply cut off as many holes as you need. The 2.0mm plates are available as cuttable/malleable versions and function as easily contoured reconstruction plates. 2.7mm Reconstruction Plates. Specifically designed for easy 3 dimensional contouring (not more than 15"). The longer plates can be cut to size. Works well for pelvic fractures and other non-weight bearing fractures. Holes: 7 Length in mm: 56
Catalog No. 50-005-82780 Supplier Med Vet International Supplier No. JOR374PS
JorVet is pleased to introduce a full range of cuttable plates from 1.5mm through to 3.5mm. The holes are round and spaced to offer the maximum number of screws per bone fragment. Plates are contoured longitudinally for extra strength. Simply cut off as many holes as you need. The 2.0mm plates are available as cuttable/malleable versions and function as easily contoured reconstruction plates. 2.7mm Reconstruction Plates. Specifically designed for easy 3 dimensional contouring (not more than 15"). The longer plates can be cut to size. Works well for pelvic fractures and other non-weight bearing fractures. Holes: 7 Length in mm: 56