Learn More
Agilent Technologies RECOVEREASE DNA ISOLATION KIT
Catalog No. 50126011
Encompass
RecoverEase DNA Isolation Kit recovers high-molecular-weight (100 to 500 kb) genomic DNA from whole tissue. It is ideal for DNA isolation for transgenic mutation assays.No organic extractions or ethanol precipitations are required.
Catalog No. 50-126-011 Supplier Agilent Technologies Supplier No. 720203
RecoverEase DNA Isolation Kit recovers high-molecular-weight (100 to 500 kb) genomic DNA from whole tissue. It is ideal for DNA isolation for transgenic mutation assays.No organic extractions or ethanol precipitations are required.