Learn More
Analytical Sales & Services Analytical Sales Recovery Vial Kit, Maximum Recovery, Pre-Slit PTFE/Silicone Liners, 100 Units
Catalog No. NC2599199
Encompass
The Maximum Recovery Vial Kit is designed for scientific applications requiring high sample recovery. With 100 clear, 1.2 mL tear-drop screw vials, each paired with pre-slit PTFE/silicone liners, it offers excellent chemical compatibility and minimizes sample loss. This kit differentiates itself with its high-quality materials and precision design, optimal for minimizing contamination and maximizing recovery in lab settings.
Catalog No. NC2599199 Supplier Analytical Sales & Services Supplier No. 87878SL
The Maximum Recovery Vial Kit is designed for scientific applications requiring high sample recovery. With 100 clear, 1.2 mL tear-drop screw vials, each paired with pre-slit PTFE/silicone liners, it offers excellent chemical compatibility and minimizes sample loss. This kit differentiates itself with its high-quality materials and precision design, optimal for minimizing contamination and maximizing recovery in lab settings.