Learn More
Aviva Systems Biology Corp RECQL5 Antibody - middle region (ARP36360_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36360_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RECQL5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RECQL5. Alternate names: FLJ90603, RECQ5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56126