Learn More
Electron Microscopy Sciences Recessed SEM Clamp, 25 x 15 mm, 3.2 mm
SDP
Catalog No. 501927145
Encompass_Preferred
Achieve background-free imaging and no contact with sample surface in imaging area with this 5 mm recessed clip. Clamping area is 25 mm (1") long x 15 mm (0.6") wide. Maximum thickness: 7 mm under the clamping strips. Made of machined aluminum with brass screws.
Size: 25 x 15 mm
Pin diameter: 3.2 mm
Catalog No. 50-192-7145 Supplier Electron Microscopy Sciences Supplier No. 7595405
Achieve background-free imaging and no contact with sample surface in imaging area with this 5 mm recessed clip. Clamping area is 25 mm (1") long x 15 mm (0.6") wide. Maximum thickness: 7 mm under the clamping strips. Made of machined aluminum with brass screws.
Size: 25 x 15 mm
Pin diameter: 3.2 mm