Learn More
Starna Cells Inc RECT.FLUOROMETER 10MM PATHLINK
SDP
Catalog No. NC9808499
Encompass_Preferred
Fluorometer Cells Standard Rectangular, Type 3, Window and Base Thickness: 1.25mm, Polished Windows: 4, Pathlength: 10mm, Window material: Optical Glass, Useable range: 334 to 2500nm, Nominal Volume: 3.5 mL, Exterior: 12.5L x 12.5W x 45mmH,
Catalog No. NC9808499 Supplier Starna Cells Inc Supplier No. 3G10
Fluorometer Cells Standard Rectangular, Type 3, Window and Base Thickness: 1.25mm, Polished Windows: 4, Pathlength: 10mm, Window material: Optical Glass, Useable range: 334 to 2500nm, Nominal Volume: 3.5 mL, Exterior: 12.5L x 12.5W x 45mmH,