Learn More
Hach Company Rect Storage Bottle PE 250 mL
Supplier:  Hach Company 2086776
Encompass
Bottle-Storage-PE-Rectangular-250 mL-12/pkHeavy-walled high-density polyethylene (HDPE) bottle features wide mouth for easy filling and leak-proof polypropylene cap Rectangular shape for maximum space efficiency and less damage in shipping Suitable for both aqueous solutions and solid samples
Catalog No. 50-002-07740