Learn More
Physitemp RET-3 Rectal Probe for Mice
Catalog No. NC9713069
Encompass_Preferred
Probe; Rectal; Animal; Rectal probe for mice; Smooth ball tip; Stainless steel shaft; 3/4in. long; 028in. dia.; Tip diameter .065in.; Time constant 0.5 secs; Not Isolated; For fast intermittent measurements
Catalog No. NC9713069 Supplier Physitemp Supplier No. RET3
Probe; Rectal; Animal; Rectal probe for mice; Smooth ball tip; Stainless steel shaft; 3/4in. long; 028in. dia.; Tip diameter .065in.; Time constant 0.5 secs; Not Isolated; For fast intermittent measurements