Learn More
Braintree Scientific Rectal Probe for Neonatal Mice, o.d. 0.020""""
SDP
Catalog No. 501955661
Encompass
Animal Rectal Probes for fast intermittent measurements. Smooth ball tip. Stainless steel shaft. Smooth ball tip, 1" long stainless steel shaft, .059" dia. .125" tip diameter, 5' lead length. Time constant .8 secs. The RET-4 Rectal probe for very small animals (neonatal mice). Flexible polyurethane sheath with smooth rounded tip (see inset). 36" long (can be made longer). Outside diameter nominal 0.032" (max at tip 0.04"). Time constant 0.18 seconds. Max temp. 140C. May be sterilized by Gas (ETO), Gluteraldehyde solution or autoclave. RET-5 rectal probe for very small animals (neonatal mice). Flexible polyurethane sheath with smooth rounded tip. 36 long (can be made longer). Outside diameter nominal 0.020. Time constant 0.1 seconds. Max temp. 140C. May be sterilized by Gas (ETO), Gluteraldehyde solution. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5661 Supplier Braintree Scientific Supplier No. RET5
Animal Rectal Probes for fast intermittent measurements. Smooth ball tip. Stainless steel shaft. Smooth ball tip, 1" long stainless steel shaft, .059" dia. .125" tip diameter, 5' lead length. Time constant .8 secs. The RET-4 Rectal probe for very small animals (neonatal mice). Flexible polyurethane sheath with smooth rounded tip (see inset). 36" long (can be made longer). Outside diameter nominal 0.032" (max at tip 0.04"). Time constant 0.18 seconds. Max temp. 140C. May be sterilized by Gas (ETO), Gluteraldehyde solution or autoclave. RET-5 rectal probe for very small animals (neonatal mice). Flexible polyurethane sheath with smooth rounded tip. 36 long (can be made longer). Outside diameter nominal 0.020. Time constant 0.1 seconds. Max temp. 140C. May be sterilized by Gas (ETO), Gluteraldehyde solution. **Sold for research purposes only and not for human use**