Learn More
Braintree Scientific Rectal Probe for Mice, 3/4"""" L .028dia. .065 tip
SDP
Catalog No. NC9656553
Encompass
Animal Rectal Probes for fast intermittent measurements. Smooth ball tip. Stainless steel shaft. Smooth ball tip, 1" long stainless steel shaft, .059" dia. .125" tip diameter, 5' lead length. Time constant .8 secs. The RET-4 Rectal probe for very small animals (neonatal mice). Flexible polyurethane sheath with smooth rounded tip (see inset). 36" long (can be made longer). Outside diameter nominal 0.032" (max at tip 0.04"). Time constant 0.18 seconds. Max temp. 140C. May be sterilized by Gas (ETO), Gluteraldehyde solution or autoclave. **Sold for research purposes only and not for human use**
Catalog No. NC9656553 Supplier Braintree Scientific Supplier No. RET3
Animal Rectal Probes for fast intermittent measurements. Smooth ball tip. Stainless steel shaft. Smooth ball tip, 1" long stainless steel shaft, .059" dia. .125" tip diameter, 5' lead length. Time constant .8 secs. The RET-4 Rectal probe for very small animals (neonatal mice). Flexible polyurethane sheath with smooth rounded tip (see inset). 36" long (can be made longer). Outside diameter nominal 0.032" (max at tip 0.04"). Time constant 0.18 seconds. Max temp. 140C. May be sterilized by Gas (ETO), Gluteraldehyde solution or autoclave. **Sold for research purposes only and not for human use**