Learn More
World Precision Instrument Rectal Temperature Probe, 1.2mm shaft diameter, 2.5mm ball
SDP
Supplier:  World Precision Instrument 61824
Encompass
Probe Sterilization Methods
The needle microprobes and animal rectal probes are all designed to withstand sterilization by ETO gas and gluteraldehyde solution. Our flexible implantable microprobes are both ETO gas sterilizable and autoclavable.
Catalog No. 50-252-1239