Learn More
Alkali Scientific Rectangle Cover Glass, Cover Slip for Microscope Slides, 24X50mm, No. 1.5 Thickness 0.17-0.19mm, 10 oz 1000/cs
SDP
Catalog No. 502389150
Encompass_Preferred
ASI™ Rectangle Cover Glass are high quality imported glass, and are non-cytotoxic. They are manufactured in 100,000 grade clean room and resistant to organic solvents and autoclaved by high temperature.
Catalog No. 50-238-9150 Supplier Alkali Scientific Supplier No. SM2107C
ASI™ Rectangle Cover Glass are high quality imported glass, and are non-cytotoxic. They are manufactured in 100,000 grade clean room and resistant to organic solvents and autoclaved by high temperature.