Learn More
Bellco Glass, Inc. Rectangular Cover Slip,10.5x35mm #1 Thick
SDP
Catalog No. 501944684
Encompass_Preferred
Glass COVER SLIP Ideal for neutral tissue cell cultures and hard to attach cells. Specifically developed for fluorescence and confocal microscopy.. Provides quality, reliability, and performance for all laboratory microscopy needs. #1 thickness (0.13 to 0.16)
Catalog No. 50-194-4684 Supplier Bellco Glass, Inc. Supplier No. 191610535A
Glass COVER SLIP Ideal for neutral tissue cell cultures and hard to attach cells. Specifically developed for fluorescence and confocal microscopy.. Provides quality, reliability, and performance for all laboratory microscopy needs. #1 thickness (0.13 to 0.16)