Learn More
Bellco Glass, Inc. Retangular Cover Slip,10.5x50mm, #1 Thick
SDP
Catalog No. 501944685
Encompass_Preferred
Glass COVER SLIP Ideal for neutral tissue cell cultures and hard to attach cells. Specifically developed for fluorescence and confocal microscopy.. Provides quality, reliability, and performance for all laboratory microscopy needs. #1 thickness (0.13 to 0.16)
Catalog No. 50-194-4685 Supplier Bellco Glass, Inc. Supplier No. 191610550A
Glass COVER SLIP Ideal for neutral tissue cell cultures and hard to attach cells. Specifically developed for fluorescence and confocal microscopy.. Provides quality, reliability, and performance for all laboratory microscopy needs. #1 thickness (0.13 to 0.16)