Learn More
Perkin Elmer US LLC Quartz SUPRASIL Macro Cell with Lid - 10 mm, 3.5 mL
Catalog No. 509051777
Encompass_Preferred
Standard rectangular macro cells are the most frequently-used type of spectroscopy cell for routine liquids analysis.
Catalog No. 50-905-1777 Supplier Perkin Elmer US LLC Supplier No. B0631107
Standard rectangular macro cells are the most frequently-used type of spectroscopy cell for routine liquids analysis.