Learn More
Thomas Scientific Rectangular cover glass, #1.5 thickness, 22 x 30 mm, 50 pack
SDP
Supplier:  Thomas Scientific 1217N86
Encompass_Preferred
High quality coverslips glasses are essential for microscopy and imaging. Cover glasses with diameters of 5 mm and 8 mm are now available from stock in convenient packages of 100.
Catalog No. NC1272771