Learn More
Thomas Scientific Rectangular cover glass, #1.5 thickness, 22 x 40 mm, 50 pack
SDP
Catalog No. NC1272772
Encompass_Preferred
High quality coverslips glasses are essential for microscopy and imaging. Cover glasses with diameters of 5 mm and 8 mm are now available from stock in convenient packages of 100.
Catalog No. NC1272772 Supplier Thomas Scientific Supplier No. 1217N87
High quality coverslips glasses are essential for microscopy and imaging. Cover glasses with diameters of 5 mm and 8 mm are now available from stock in convenient packages of 100.