Learn More
Electron Microscopy Sciences 5" Shaft, Rectangular Foam Tip Swab 11/16"W x ⅞"L, Polypropylene Shaft, 50/pk
SDP
Catalog No. 50949232
Encompass_Preferred
Tips withstand many chemicals and solutions used in labs and clean rooms. Resistant to deterioration. Head is made from 100% urethane foam. Foam-tip on a 5" polypropylene shaft, head size: 1-1/16".
Catalog No. 50-949-232 Supplier Electron Microscopy Sciences Supplier No. 72314
Tips withstand many chemicals and solutions used in labs and clean rooms. Resistant to deterioration. Head is made from 100% urethane foam. Foam-tip on a 5" polypropylene shaft, head size: 1-1/16".