Learn More
MSC Value Collection Rectangular Steel Gage Block, 0.142 in., Grade AS-1, 1 ea.
Catalog No. 5000510790
Encompass
This rectangular steel gage block is designed for precision measurement applications and features an accuracy grade of AS-1. Each unit is manufactured to maintain high dimensional stability and reliability for inspection and calibration tasks.
  • Accuracy grade AS-1
  • Constructed from steel
  • Includes NIST traceability certification
  • Size: 0.142 in.
  • Made in the USA
Catalog No. 50-005-10790 Supplier MSC Supplier No. 86293107
This rectangular steel gage block is designed for precision measurement applications and features an accuracy grade of AS-1. Each unit is manufactured to maintain high dimensional stability and reliability for inspection and calibration tasks.
  • Accuracy grade AS-1
  • Constructed from steel
  • Includes NIST traceability certification
  • Size: 0.142 in.
  • Made in the USA