Learn More
MSC Value Collection Rectangular Gage Block, Steel, 0.129 in., Grade 0, 1 ea.
Catalog No. 5000510739
Encompass
This rectangular gage block is designed for precision measurement and calibration applications. Constructed from steel, it provides a stable and accurate reference for machine setup and inspection tasks.
  • Accuracy grade 0
  • Rectangular shape
  • Steel material
  • Includes NIST traceability certification
Catalog No. 50-005-10739 Supplier MSC Supplier No. 86282456
This rectangular gage block is designed for precision measurement and calibration applications. Constructed from steel, it provides a stable and accurate reference for machine setup and inspection tasks.
  • Accuracy grade 0
  • Rectangular shape
  • Steel material
  • Includes NIST traceability certification