Learn More
MSC Value Collection Rectangular Steel Gage Block, 0.1008 in., AS-1, 1 ea.
Catalog No. 5000510730
Encompass
This rectangular steel gage block is designed for precise measurement and calibration applications. Featuring a dimension of 0.1008 in. and manufactured to Grade AS-1 accuracy standards, this block provides reliable performance for high-precision metrology tasks.
  • Manufactured from durable steel
  • Grade AS-1 accuracy for high precision
  • Includes NIST traceability certification
  • Made in the USA
Catalog No. 50-005-10730 Supplier MSC Supplier No. 86291085
This rectangular steel gage block is designed for precise measurement and calibration applications. Featuring a dimension of 0.1008 in. and manufactured to Grade AS-1 accuracy standards, this block provides reliable performance for high-precision metrology tasks.
  • Manufactured from durable steel
  • Grade AS-1 accuracy for high precision
  • Includes NIST traceability certification
  • Made in the USA