Learn More
MSC Value Collection Rectangle Steel Gage Block, 0.106 in., Grade AS-1, 1 ea.
Catalog No. 5000510650
Encompass
This rectangle steel gage block provides a precise reference standard for dimensional measurement and calibration tasks. Manufactured for accuracy and stability, it is suitable for use in industrial metrology and quality control environments.
  • Accuracy grade AS-1
  • Constructed from steel
  • Rectangle shape
  • Includes NIST traceability certification
  • Size of 0.1060 in.
Catalog No. 50-005-10650 Supplier MSC Supplier No. 86291358
This rectangle steel gage block provides a precise reference standard for dimensional measurement and calibration tasks. Manufactured for accuracy and stability, it is suitable for use in industrial metrology and quality control environments.
  • Accuracy grade AS-1
  • Constructed from steel
  • Rectangle shape
  • Includes NIST traceability certification
  • Size of 0.1060 in.