Learn More
MSC Mitutoyo Rectangle Gage Block, Steel, 0.21 in., Grade 0, 1 ea.
Catalog No. 5000510303
Encompass
This rectangular steel gage block is designed for precise calibration and inspection applications. Crafted from high-carbon, high-chrome steel, it ensures high hardness and exceptional wear resistance for reliable performance in measurement and machine setup tasks.
  • Accuracy grade 0 for precision measurement
  • Rectangular base facilitates accurate wringing
  • Etched identification number ensures traceability
  • Includes certificate of inspection
  • Meets ASME standards
Catalog No. 50-005-10303 Supplier MSC Supplier No. 88608831
This rectangular steel gage block is designed for precise calibration and inspection applications. Crafted from high-carbon, high-chrome steel, it ensures high hardness and exceptional wear resistance for reliable performance in measurement and machine setup tasks.
  • Accuracy grade 0 for precision measurement
  • Rectangular base facilitates accurate wringing
  • Etched identification number ensures traceability
  • Includes certificate of inspection
  • Meets ASME standards