Learn More
MSC Mitutoyo Rectangle Gage Block, Steel, 0.132 in., Grade 0, 1 ea.
Catalog No. 5000510278
Encompass
This individual rectangle steel gage block provides a precision reference for calibration and measurement applications. Engineered for high accuracy, it meets ASME standards and features a rectangular base design that facilitates reliable wringing. Each block includes a certificate of inspection and has an identification number etched into the surface to support traceability.
  • Constructed from high-quality steel
  • Meets ASME grade 0 accuracy standards
  • Rectangular base supports accurate wringing
  • Individual presentation
  • Identification number etched for traceability
  • Includes certificate of inspection
Catalog No. 50-005-10278 Supplier MSC Supplier No. 86668712
This individual rectangle steel gage block provides a precision reference for calibration and measurement applications. Engineered for high accuracy, it meets ASME standards and features a rectangular base design that facilitates reliable wringing. Each block includes a certificate of inspection and has an identification number etched into the surface to support traceability.
  • Constructed from high-quality steel
  • Meets ASME grade 0 accuracy standards
  • Rectangular base supports accurate wringing
  • Individual presentation
  • Identification number etched for traceability
  • Includes certificate of inspection