Learn More
MSC Value Collection Rectangular Steel Gage Block, 0.147 in., Grade AS-1, 1 ea
Catalog No. 5000515415
Encompass
This rectangular steel gage block is designed for precision measurement, layout, and machine setup applications. Engineered for durability and reliability, it serves as a high-quality standard for calibrating inspection equipment in industrial settings.
  • Manufactured from high-grade steel
  • Rectangular shape for versatile use
  • AS-1 accuracy grade
  • Includes NIST traceability certification
Catalog No. 50-005-15415 Supplier MSC Supplier No. 86293354
This rectangular steel gage block is designed for precision measurement, layout, and machine setup applications. Engineered for durability and reliability, it serves as a high-quality standard for calibrating inspection equipment in industrial settings.
  • Manufactured from high-grade steel
  • Rectangular shape for versatile use
  • AS-1 accuracy grade
  • Includes NIST traceability certification