Learn More
MSC Value Collection Rectangle Steel Gage Block, 0.5 in., Grade AS-1, NIST Certified, 1 ea.
Catalog No. 5000515085
Encompass
This rectangle steel gage block is designed for precision measurement applications and features a nominal size of 0.5 in. with a grade AS-1 tolerance level.
  • Constructed from high-quality steel for durability and stability
  • Includes official NIST certification to ensure traceability and accuracy
  • Rectangle geometry suitable for a variety of calibration and inspection tasks
  • Nominal size of 0.5000 in.
Catalog No. 50-005-15085 Supplier MSC Supplier No. 86294709
This rectangle steel gage block is designed for precision measurement applications and features a nominal size of 0.5 in. with a grade AS-1 tolerance level.
  • Constructed from high-quality steel for durability and stability
  • Includes official NIST certification to ensure traceability and accuracy
  • Rectangle geometry suitable for a variety of calibration and inspection tasks
  • Nominal size of 0.5000 in.