Learn More
MSC Mitutoyo Rectangular Gage Block, Steel, 0.55 in., Grade 0, 1 ea.
Catalog No. 5000511810
Encompass
This rectangle-shaped steel gage block provides a precise 0.55 in. reference standard for calibration and dimensional measurement applications. Designed to meet ASME Grade 0 standards, this individual mechanical gage block is manufactured to ensure high accuracy in layout and machine setup tasks.
  • Individual 0.55 in. rectangular steel gage block
  • Grade 0 accuracy for high-precision applications
  • Repeated heat treatment process to minimize dimensional change over time
  • Includes a certificate of inspection
  • Meets ASME standards
Catalog No. 50-005-11810 Supplier MSC Supplier No. 86707718
This rectangle-shaped steel gage block provides a precise 0.55 in. reference standard for calibration and dimensional measurement applications. Designed to meet ASME Grade 0 standards, this individual mechanical gage block is manufactured to ensure high accuracy in layout and machine setup tasks.
  • Individual 0.55 in. rectangular steel gage block
  • Grade 0 accuracy for high-precision applications
  • Repeated heat treatment process to minimize dimensional change over time
  • Includes a certificate of inspection
  • Meets ASME standards