Learn More
MSC Value Collection Rectangle Gage Block, Steel, 0.114 in., Grade 0, 1 ea.
Catalog No. 5000511797
Encompass
This rectangular steel gage block is designed for precise calibration and inspection applications. Crafted for dimensional accuracy, it serves as a reliable standard in measurement and machine setup processes.
  • Constructed from durable steel
  • Grade 0 accuracy for high-precision requirements
  • Rectangular shape for versatile use
  • NIST certification ensures standard traceability
  • Size of 0.1140 in.
Catalog No. 50-005-11797 Supplier MSC Supplier No. 86281755
This rectangular steel gage block is designed for precise calibration and inspection applications. Crafted for dimensional accuracy, it serves as a reliable standard in measurement and machine setup processes.
  • Constructed from durable steel
  • Grade 0 accuracy for high-precision requirements
  • Rectangular shape for versatile use
  • NIST certification ensures standard traceability
  • Size of 0.1140 in.