Learn More
MSC Value Collection Rectangle Steel Gage Block, 5 in., Grade 0, 1 ea.
Catalog No. 5000511801
Encompass
This rectangular steel gage block is designed for precision measurement and calibration applications. Featuring a 5 in. length, this tool provides reliable accuracy for layout and machine setup tasks in industrial environments.
  • Manufactured from high-quality steel
  • Grade 0 accuracy for precise measurements
  • NIST traceability certification included
  • Rectangular shape for versatile use
Catalog No. 50-005-11801 Supplier MSC Supplier No. 86286101
This rectangular steel gage block is designed for precision measurement and calibration applications. Featuring a 5 in. length, this tool provides reliable accuracy for layout and machine setup tasks in industrial environments.
  • Manufactured from high-quality steel
  • Grade 0 accuracy for precise measurements
  • NIST traceability certification included
  • Rectangular shape for versatile use