Learn More
MSC Mitutoyo Rectangular Steel Gage Block, 0.145 in., ASME Grade AS-1, 1 ea
Catalog No. 5000511696
Encompass
The rectangular steel gage block is designed for high-precision metrology and calibration applications. This individual gage block provides reliable and stable measurement standards for industrial inspection tasks.
  • ASME grade AS-1 accuracy
  • Constructed from high-carbon, high-chrome steel
  • High degree of hardness for durability
  • Laser-inscribed characters
  • Includes certificate of inspection
  • Registered deviation for each block
Catalog No. 50-005-11696 Supplier MSC Supplier No. 88609359
The rectangular steel gage block is designed for high-precision metrology and calibration applications. This individual gage block provides reliable and stable measurement standards for industrial inspection tasks.
  • ASME grade AS-1 accuracy
  • Constructed from high-carbon, high-chrome steel
  • High degree of hardness for durability
  • Laser-inscribed characters
  • Includes certificate of inspection
  • Registered deviation for each block