Learn More
Cole-Parmer LDPE Rectangular Mobile Tub with Lid, 15 gal; 1/Pk
Supplier:  Cole-Parmer 0674520
Encompass_Preferred
Tubs are made of low-density polyethylene (LDPE). Rugged caster frame supports heavy loads. Three-inch diameter casters make rolling easy. Container and casters can be removed from zinc-plated frame for cleaning. Tubs include polyethylene cover. NSF certified; USDA approved.
Catalog No. NC9613195