Learn More
Electron Microscopy Sciences Rectangular Stain Dish with glass cover
SDP
Catalog No. 50363964
Encompass_Preferred
Heavy, clear, soda lime glass stain dish, accept glass slide measure 75 x 25 mm or 3 x 1" . This dish comes with glass cover, holds 20 slides back-to-back horizontally. Inside dimensions: 3" x 2 1/4"x 1 1/4 deep.
Catalog No. 50-363-964 Supplier Electron Microscopy Sciences Supplier No. 7031401
Heavy, clear, soda lime glass stain dish, accept glass slide measure 75 x 25 mm or 3 x 1" . This dish comes with glass cover, holds 20 slides back-to-back horizontally. Inside dimensions: 3" x 2 1/4"x 1 1/4 deep.