Learn More
Perkin Elmer US LLC Semi-Demountable Cell Window - ZnSe, 2 mm, Qty. 2
Catalog No. 509043312
Encompass_Preferred
Semi-Demountable Cell Windows are rectangular windows and come in a variety of window materials. Each window measures 41 (H) x 23 mm (W), and they come in pairs. These windows require, but do not include, the Universal Demountable Cell (L1270986).
Catalog No. 50-904-3312 Supplier Perkin Elmer US LLC Supplier No. L1270765
Semi-Demountable Cell Windows are rectangular windows and come in a variety of window materials. Each window measures 41 (H) x 23 mm (W), and they come in pairs. These windows require, but do not include, the Universal Demountable Cell (L1270986).