Learn More
MSC Delfin Industrial Rectangular Brush, FDA Certified, Autoclavable, NBR/Nylon, 1/Each
Catalog No. 502815383
Encompass
This rectangular brush is FDA certified and autoclavable, designed for cleaning flat, ground, or overhead surfaces in pharmaceutical and food industries. It features nylon bristles and a 2 in wide opening for attachment to a vacuum hose, providing versatility for various cleaning tasks.
  • For use with 2 in hose cuff vacuums
  • Ideal for ground or high-up surface cleaning
  • Rectangular shape of 6 1/2 × 3 1/2 in
  • NBR/Nylon construction
  • Green color for contamination control through color-coding
  • Connects directly to 50 mm hoses; requires adapter for 40 mm hoses
  • Compliant with 652 standards
Catalog No. 50-281-5383 Supplier MSC Supplier No. 13676911
This rectangular brush is FDA certified and autoclavable, designed for cleaning flat, ground, or overhead surfaces in pharmaceutical and food industries. It features nylon bristles and a 2 in wide opening for attachment to a vacuum hose, providing versatility for various cleaning tasks.
  • For use with 2 in hose cuff vacuums
  • Ideal for ground or high-up surface cleaning
  • Rectangular shape of 6 1/2 × 3 1/2 in
  • NBR/Nylon construction
  • Green color for contamination control through color-coding
  • Connects directly to 50 mm hoses; requires adapter for 40 mm hoses
  • Compliant with 652 standards