Learn More
Alkali Scientific Rectangle Cover Glass, Cover Slip, for Microscope Slides, 24 X 60mm, No. 1, Thickness 0.13 to 0.17mm, 1 oz 100/Sleeve
SDP
Catalog No. 502335816
Encompass_Preferred
ASI™ Rectangle Cover Glass are high quality imported glass, and are non-cytotoxic. They are manufactured in 100,000 grade clean room and resistant to organic solvents and autoclaved by high temperature.
Catalog No. 50-233-5816 Supplier Alkali Scientific Supplier No. SM2460
ASI™ Rectangle Cover Glass are high quality imported glass, and are non-cytotoxic. They are manufactured in 100,000 grade clean room and resistant to organic solvents and autoclaved by high temperature.