Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Recykleen™ Sharps Collection Trolley

Catalog No. 22204436 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Capacity (English):
17 to 19 gal.
9 gal.
Product Type:
Foot-operated Trolley
Standard Trolley
Height (Metric):
59.6 cm
61.2 cm
Width (Metric):
41.4 cm
45.7 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Product Type Height (Metric) Width (Metric)
22-204-436 17 to 19 gal. Standard Trolley 61.2 cm 45.7 cm
22-204-435 9 gal. Foot-operated Trolley 59.6 cm 41.4 cm
22-204-434 9 gal. Standard Trolley 59.6 cm 41.4 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-204-436 Supplier BD Supplier No. 305092
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

These collectors are made from a minimum 70% recycled material and are intended for Resource Conservation and Recovery Act (RCRA) solid hazardous waste. As an added feature, each RCRA collector comes with a liquid-absorbent material preattached to the inside bottom of the collector.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 64.4 to 71.9 L
Depth (English) 27.5 in.
Depth (Metric) 69.8 cm
Capacity (English) 17 to 19 gal.
Color Gray
Height (Metric) 61.2 cm
Sterility Non-sterile
Width (Metric) 45.7 cm
Includes Liquid-absorbent material
For Use With (Application) Design that reduces the risk of collectors tipping over
For Use With (Equipment) For all 17 and 19 gal.
Width (English) 18 in.
Height (English) 24.1 in.
Material Recycled Material
Product Type Standard Trolley
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.