Learn More
Thomson Instrument Company Red 11mm Snap Cap, Pre-Slit Septa
SDP
Catalog No. NC2339421
Encompass_Preferred
Thomson Instrument Company’s Snap Caps used with Thomson’s vials which are universal and can be used on most autosamplers. Thomson manufactures its products in the USA to assure same day shipping.
Catalog No. NC2339421 Supplier Thomson Instrument Company Supplier No. 70652R2
Thomson Instrument Company’s Snap Caps used with Thomson’s vials which are universal and can be used on most autosamplers. Thomson manufactures its products in the USA to assure same day shipping.