Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Red 2G, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014077 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4077 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4077 Supplier MilliporeSigma Supelco Supplier No. 4046225MG
Add to Cart
Add to Cart

Red 2G is a compound, which belongs to the azo family of dyes, generally used in textile industry.It can mainly find applications in non-linear optical studies.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 3734-67-6
Physical Form Neat
Linear Formula C18H13N3 Na2O8S2
MDL Number MFCD00003954
Synonym Acid Red 1; Amido Naphthol Red G; Azophloxine
Type Analytical Standard
Quantity 25 mg
Formula Weight 509.42
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.