Learn More
Cytek Biosciences Inc RED 710 MS CD45.2 (104) 100UG
Supplier:  Cytek Biosciences Inc 800454U100
Encompass_Preferred
redFluor 710 Anti-Mouse CD45.2 (104) Isotype: Mouse IgG2a, kappa Apps: FC Size: 100 ug
Catalog No. 50-105-5218