Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Red-Al™ Sodium Bis(2-Methoxyethoxy)Aluminum Hydride Solution

Catalog No. 196193500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-619-3500G 500 g
19-619-350G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 19-619-3500G Supplier Sigma Aldrich Supplier No. 196193500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Red-Al™ sodium bis(2-methoxyethoxy)aluminum hydride [NaAlH2(OCH2CH2OCH3)2] is used as a versatile reducing agent in organic synthesis. It is used to reduce: Aldehydes, ketones, esters, and anhydrides to primary alcohols. Ketoximes and aldoximes to primary amines. Cyclic compounds such as lactones and epoxides to diols.It is also used as a catalyst in the ring-opening polymerization reactions.Red-Al™ sodium bis(2-methoxyethoxy)aluminum hydride can be used as: A tosyl deprotecting agent A catalyst for crosslinking of polyvinylsilanes (PVS) by Si-Si dehydrocoupling reaction in the presence of group 4 metallocene complexes A reducing reagent for the synthesis of optically active N-protected amino alcohols and peptide alcohols.Red-Al is a registered trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 22722-98-1
Density 1.036 g/mL at 25°C
Molecular Formula C6H16AlNaO4
Linear Formula NaAlH2(OCH2CH2OCH3)2
MDL Number MFCD00011631
Quantity 500 g
Synonym SBAH; Sodium bis(2-methoxyethoxy)aluminum dihydride; Sodium dihydrido-bis(2-methoxyethoxy)aluminate
Molecular Weight (g/mol) 202.16 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.