Learn More
Montana Molecular Alpha Tubulin-Targeted Montana Paintbrush (Red AlphaTubulinPaint) Expression Kit - Big Sky BacMam, 5mL - Montana Molecular
SDP
Catalog No. NC3787559
Encompass_Preferred
Bright, red fluorescent protein fused to alpha tubulin for live-cell imaging—no fixation, staining, or cell lysis required. Visualize cytoskeletal structures and cell morphology, over hours to days, in living cells. Delivered using Montana Molecular’s Big Sky BacMam vector for enhanced expression across a wide range of cell types, including primary cultures and iPSCs. Ideal for non-destructive studies of alpha tubulin organization, cellular phenotypes, and high-content imaging.
Catalog No. NC3787559 Supplier Montana Molecular Supplier No. P1070R005
Bright, red fluorescent protein fused to alpha tubulin for live-cell imaging—no fixation, staining, or cell lysis required. Visualize cytoskeletal structures and cell morphology, over hours to days, in living cells. Delivered using Montana Molecular’s Big Sky BacMam vector for enhanced expression across a wide range of cell types, including primary cultures and iPSCs. Ideal for non-destructive studies of alpha tubulin organization, cellular phenotypes, and high-content imaging.