Learn More
Research Products International Corp Red Biohazard Bags, 8.5x11"", 100/cs
SDP
Catalog No. 502252975
Encompass
Bright red, 2.25 mil thick autoclavable bags are designed for easy identification of biohazardous waste. Made from durable polypropylene, these bags feature a printed biohazard symbol and a sterilization indicator patch, ensuring safe handling and disposal. Their robust construction provides added safety, making them ideal for managing biohazardous materials with confidence.
This item is not returnable. View return policy
Catalog No. 50-225-2975 Supplier Research Products International Corp Supplier No. 248522
Bright red, 2.25 mil thick autoclavable bags are designed for easy identification of biohazardous waste. Made from durable polypropylene, these bags feature a printed biohazard symbol and a sterilization indicator patch, ensuring safe handling and disposal. Their robust construction provides added safety, making them ideal for managing biohazardous materials with confidence.