Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Red Blood Cells Rabbit anti-Sheep, Polyclonal, MP Biomedicals™

Catalog No. MP0855800 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP0855800 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP0855800 Supplier MP Biomedicals, Inc Supplier No. 0855800
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

  • Rabbit polyclonal to Sheep Red Blood Cell Stroma.
  • Red blood cells polyclonal antibody is used in immunoblotting; Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Red blood cells
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal to Sheep Red Blood Cell Stroma.
Formulation 0.02M sodium phosphate, 0.14M sodium chloride, pH 7.3
Host Species Rabbit
Purification Method solid-phase absorption
Quantity 2 mL
Regulatory Status RUO
Primary or Secondary Primary
Reconstitution Reconstitute product with 2.0mL of deionized or distilled water.gentle swirling may be used to speed rehydration. Avoid vigorous shaking of the reconstituted material.
Target Species Sheep
Content And Storage Store lyophilized product at 2-8°C. Store reconstituted material no longer than 4 weeks at 2-8°C. For long-term storage, aliquot and freeze reconstituted product at -20°C or lower.
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.