Learn More
AdipoGen Red C2-dichlorotriazine
SDP
Supplier:  AdipoGen CDXS0077M001
Encompass_Preferred
Chemical. Formula C36H35Cl2N7O6S2. MW 796.74. Synthetic. Amine-reactive dye that modifies amines in proteins. Reported to react directly with polysaccharides and other alcohols in aqueous solution, provided that the pH is >9 and that other nucleophiles are absent. Shows absorption/emission maxima of ~588/601 nm.
Catalog No. 50-256-0069