Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Red Cabbage Juice pH Indicator
SCI_ED

Catalog No. S280005 Shop All Aldon Products
Change view
Click to view available options
pH Range:
0 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
S280005 0 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S280005 Supplier Innovating Science™ Supplier No. IS5068
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Learn about acids, bases, and pH indicators with this Red Cabbage Juice pH Indicator.

  • Learn about acids, bases, and pH indicators with our red cabbage juice kit.
  • A pH indicator is a chemical that behaves differently depending on the pH level it is exposed to.
  • Red cabbage juice is a natural pH indicator that changes color from red to purple to yellow-green as the pH of a solution increases.
  • Test out different substances to determine if they are acidic or basic, and see if you can make all six colors found on the color chart!

Specifications

Color White
pH Range 0 to 14
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.