Learn More
Perkin Elmer US LLC Red, Capillary split/splitless glass injector liner packed loosely with wool and tapered end
Catalog No. 502104652
Encompass_Preferred
Application - Focus liner for split/splitless operation. Wipes the sample from the tip of the needle for complete transfer Gives accurate and reproducible results Performs well in splitless mode Promotes vaporization of sample Minimizes mass discrimination Prevents non-volatile material entering the column (acts as a filter) Color coded RED on the liner with PKI name.
Catalog No. 50-210-4652 Supplier Perkin Elmer US LLC Supplier No. N9306235
Application - Focus liner for split/splitless operation. Wipes the sample from the tip of the needle for complete transfer Gives accurate and reproducible results Performs well in splitless mode Promotes vaporization of sample Minimizes mass discrimination Prevents non-volatile material entering the column (acts as a filter) Color coded RED on the liner with PKI name.