Learn More
Microsolv Technology Corporation Red Caps 1000Pk
Catalog No. NC1530276
Encompass_Preferred
Caps, Screw Top, with White Silicone Rubber / Natural PTFE Pre-Slit Septa, fitted in knurled, polypropylene, Red Caps. For use with 9-425mm thread, 12x32mm autosampler vials. BASIK Brand. 1000/CS.
Catalog No. NC1530276 Supplier Microsolv Technology Corporation Supplier No. 9502SE35RM
Caps, Screw Top, with White Silicone Rubber / Natural PTFE Pre-Slit Septa, fitted in knurled, polypropylene, Red Caps. For use with 9-425mm thread, 12x32mm autosampler vials. BASIK Brand. 1000/CS.